N-Boc-glycin (Boc-Gly-OH)
Lieferant: NOVABIOCHEM
Synonyme: Boc-Gly-OH
8.53000.0100 8.53000.0025
04-12-0001-100EA 95.6 EUR
04-12-0001-100 04-12-0001-25
N-Boc-glycin (Boc-Gly-OH)
N-Boc-glycin (Boc-Gly-OH)
Formel: C₇H₁₃NO₄
Molecular Weight: 175.2 g/mol
MDL: MFCD00002690
CAS-Nummer: 4530-20-5
EINECS: 224-864-5