D(-)-Diisopropyltartrat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (2S,3S)-(-)-Diisopropyl tartrate, (2S,3S)-Diisopropyl 2,3-dihydroxysuccinate, D-(-)-Tartaric acid diisopropyl ester

M-Clarity™ Qualitätsstufe = MQ100

227803-10G 227803-50G
SIAL227803-10GEA 36 EUR
SIAL227803-10G SIAL227803-50G
D(-)-Diisopropyltartrat, Sigma-Aldrich®
D(-)-Diisopropyltartrat
Formel: C₁₀H₁₈O₆
Molecular Weight: 234,25 g/mol
MDL: MFCD00008876
CAS-Nummer: 62961-64-2
EINECS: 263-771-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR