N-Phenylglycin ≥97%

Lieferant: Thermo Fisher Scientific

Synonyme: 2-Anilinoacetic acid, 2-(Phenylamino)acetic acid

A14182.09 A14182.18
A14182.09EA 41 EUR
A14182.09 A14182.18
N-Phenylglycin ≥97%
N-Phenylglycin

Purity: 97% CAS No.: 103-01-5 Molecular Formula: (C6H5)NHCH2CO2H Formula Weight: 151.17 Melting Point: 117-119° Beilstein Reference: 509838 Merck Reference: 14,7292 MDL No.: MFCD00014009

Formel: C₈H₉NO₂
Molecular Weight: 151.17 g/mol
Schmelzpunkt: 117…119 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00014009
CAS-Nummer: 103-01-5
EINECS: 203-070-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR