1-Methyl-1-phenyl-2-thioharnstoff

Lieferant: Thermo Scientific

Synonyme: N-Methyl-N-phenylthiourea

TL00838.10 TL00838.50
MAYBTL00838.10EA 0 EUR
MAYBTL00838.10 MAYBTL00838.50
1-Methyl-1-phenyl-2-thioharnstoff
1-Methyl-1-phenyl-2-thioharnstoff
Formel: C₈H₁₀N₂S
Molecular Weight: 166,24 g/mol
MDL: MFCD00086629
CAS-Nummer: 4104-75-0
EINECS: 223-877-3
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR