3,6,9-Trioxa-1,11-undecandiamin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1,11-Diamino-3,6,9-trioxaundecane

40421-100MG-F 40421-500MG-F
SIAL40421-100MG-FEA 385 EUR
SIAL40421-100MG-F SIAL40421-500MG-F
3,6,9-Trioxa-1,11-undecandiamin, Sigma-Aldrich®
3,6,9-Trioxa-1,11-undecandiamin
Formel: C₈H₂₀N₂O₃
Molecular Weight: 192,2560 g/mol
MDL: MFCD00628164
CAS-Nummer: 929-75-9
EINECS: 213-206-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR