4,7,10-Trioxa-1,13-tridecandiamin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
1,13-Diamino-4,7,10-trioxatridecane, 3,3'-((Oxybis(ethane-2,1-diyl))bis(oxy))bis(propan-1-amine)
M-Clarity™ Qualitätsstufe = MQ100
369519-100G
369519-500G
SIAL369519-100GEA
67.3
EUR
SIAL369519-100G
SIAL369519-500G
4,7,10-Trioxa-1,13-tridecandiamin, Sigma-Aldrich®
4,7,10-Trioxa-1,13-tridecandiamin
Formel:
C₁₀H₂₄N₂O₃ Molecular Weight: 220.3092 g/mol |
MDL:
MFCD00059850 CAS-Nummer: 4246-51-9 EINECS: 224-207-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...