5-Chlor-3-methyl-1-phenyl-1H-pyrazol-4-carbaldehyd

Lieferant: BIONET RESEARCH

Synonyme: 5-Chloro-3-methyl-1-phenyl-1H-pyrazole-4-carboxaldehyde

11G-910.500 11G-910.1 11G-910.5 11G-910.10
BIOT11G-910.500EA 0 EUR
BIOT11G-910.500 BIOT11G-910.1 BIOT11G-910.5 BIOT11G-910.10
5-Chlor-3-methyl-1-phenyl-1H-pyrazol-4-carbaldehyd
5-Chlor-3-methyl-1-phenyl-1H-pyrazol-4-carbaldehyd
Formel: C₉H₁₁ClN₂O
Molecular Weight: 220,65 g/mol
MDL: MFCD00100755
CAS-Nummer: 947-95-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR