4-(3,5-Di-tert-butyl-1H-pyrazol-1-yl)benzaldehyd

Lieferant: Thermo Scientific

SEW06682.250 SEW06682.1
MAYBSEW06682.250EA 0 EUR
MAYBSEW06682.250 MAYBSEW06682.1
4-(3,5-Di-tert-butyl-1H-pyrazol-1-yl)benzaldehyd
4-(3,5-Di-tert-butyl-1H-pyrazol-1-yl)benzaldehyd
Formel: C₁₈H₂₄N₂O
Molecular Weight: 284,4 g/mol
MDL: MFCD02090222
CAS-Nummer: 849066-54-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR