Methyl-vinylsulfon
Lieferant: FLUOROCHEM
001782-5G
001782-25G
001782-80G
FLUO001782-5GEA
84.9
EUR
FLUO001782-5G
FLUO001782-25G
FLUO001782-80G
Methyl-vinylsulfon
Methyl-vinylsulfon
Formel:
C₃H₆O₂S Molecular Weight: 106.15 g/mol |
MDL:
MFCD00007567 CAS-Nummer: 3680-02-2 EINECS: 222-958-0 UN: 2810 ADR: 6.1 I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...