4-(N,N-Diphenylamino)phenylboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
4-(Diphenylamino)phenylboronic acid Pinacol ester, 4-(Diphenylamino)benzeneboronic acid pinacol ester, Triphenylamine-4-boronic acid Pinacol ester, N,N-Diphenyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 2-[4-(Diphenylamino)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PN-8149-5G
PN-8149-1G
COBBPN-8149-5GEA
312
EUR
COBBPN-8149-5G
COBBPN-8149-1G
4-(N,N-Diphenylamino)phenylboronsäurepinakolester
4-(N,N-Diphenylamino)phenylboronsäurepinakolester
Formel:
C₂₄H₂₆BNO₂ Molecular Weight: 371.29 g/mol |
MDL:
MFCD13195770 CAS-Nummer: 267221-88-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...