3-Boronobenzolsulfonamidpinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzenesulphonamide
PN-2403-5G
PN-2403-1G
COBBPN-2403-5GEA
724
EUR
COBBPN-2403-5G
COBBPN-2403-1G
3-Boronobenzolsulfonamidpinakolester
3-Boronobenzolsulfonamidpinakolester
Formel:
C₁₂H₁₈BNO₄S Molecular Weight: 283,1516 g/mol |
MDL:
MFCD07781217 CAS-Nummer: 486422-08-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...