4-Boronobenzolsulfonamidpinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzenesulphonamide, 4-Sulphamoylbenzeneboronic acid, pinacol ester

PN-2402-5G PN-2402-1G
COBBPN-2402-5GEA 648 EUR
COBBPN-2402-5G COBBPN-2402-1G
4-Boronobenzolsulfonamidpinakolester
4-Boronobenzolsulfonamidpinakolester
Formel: C₁₂H₁₈BNO₄S
Molecular Weight: 283,1516 g/mol
MDL: MFCD06657806
CAS-Nummer: 214360-51-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR