N-tert-Butyl-4-boronobenzolsulfonamid pinacol ester

Lieferant: COMBI-BLOCKS

Synonyme: N-(tert-Butyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzenesulphonamide

PN-2406-5G PN-2406-1G
COBBPN-2406-5GEA 884 EUR
COBBPN-2406-5G COBBPN-2406-1G
N-tert-Butyl-4-boronobenzolsulfonamid pinacol ester
N-tert-Butyl-4-boronobenzolsulfonamid pinacol ester
Formel: C₁₆H₂₆BNO₄S
Molecular Weight: 339,2579 g/mol
MDL: MFCD06657804
CAS-Nummer: 648905-63-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR