[1,1'-Bis(diphenylphosphino)ferrocen]palladium(II)chlorid
Lieferant: COMBI-BLOCKS
Synonyme:
[1,1'-Bis(diphenylphosphino)ferrocene]dichloropalladium(II), Dichloro[1,1'-bis(diphenylphosphino)ferrocene]palladium(II), Pd(dppf)Cl2, 1,1’-Bis(diphenylphosphino)ferrocene palladium(II)dichloride
OT-0746-5G
OT-0746-25G
OT-0746-1G
COBBOT-0746-5GEA
340
EUR
COBBOT-0746-5G
COBBOT-0746-25G
COBBOT-0746-1G
[1,1'-Bis(diphenylphosphino)ferrocen]palladium(II)chlorid
[1,1'-Bis(diphenylphosphino)ferrocen]palladium(II)chlorid
Formel:
C₃₄H₂₈Cl₂P₂FePd Molecular Weight: 731.70 g/mol |
MDL:
MFCD00015757 CAS-Nummer: 72287-26-4 EINECS: 460-040-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...