[1,1'-Bis(diphenylphosphino)ferrocen]palladium(II)chlorid-Dichlormethan-Komplex
Lieferant: Thermo Fisher Scientific
Achtung
Synonyme: Dichloro 1,1'-bis(diphenylphosphino)ferrocene palladium (II) dichloromethane, [1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) Dichloride Dichloromethane Adduct (1:1), Dichloro[1,1'-bis(diphenylphosphino)ferrocene]palladium(II), complex with dichloromethane, 1,1'-Bis(diphenylphosphino)ferrocene-palladium(II)dichloride dichloromethane adduct, 1,1'-Bis(diphenylphosphino)ferrocene-palladium(II)dichloride dichloromethane complex
348680010 348680050 348680250
ACRO348680010EA 106 EUR
ACRO348680010 ACRO348680050 ACRO348680250
[1,1'-Bis(diphenylphosphino)ferrocen]palladium(II)chlorid-Dichlormethan-Komplex
[1,1'-Bis(diphenylphosphino)ferrocen]palladium(II)chlorid-Dichlormethan-Komplex
Formel: C₃₅H₃₀Cl₄FeP₂Pd
Molecular Weight: 816.64 g/mol
Schmelzpunkt: 275…280 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00792899
CAS-Nummer: 95464-05-4
Bestellen
Spezifikation Testergebnisse
Appearance Orange to red-brown Crystalline powder
Infrared spectrum Conforms
Carbon (C) 50 to 53 %
Palladium (Pd) 12.6 to 13.4 %