Zink(II)phthalocyanin (Sulfat-Ionophor II), Sigma-Aldrich®

Lieferant: Merck

Synonyme: Zinc phthalocyanine, 29H,31H-Phthalocyanine, zinc complex

M-Clarity™ Qualitätsstufe = MQ100

341169-5G 341169-25G
SIAL341169-5GEA 214 EUR
SIAL341169-5G SIAL341169-25G
Zink(II)phthalocyanin (Sulfat-Ionophor II), Sigma-Aldrich®
Zink(II)phthalocyanin (Sulfat-Ionophor II)
Formel: C₃₂H₁₆N₈Zn
Molecular Weight: 577,91 g/mol
MDL: MFCD00041955
CAS-Nummer: 14320-04-8
EINECS: 238-262-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR