2-Piperidinonicotinonitril

Lieferant: BIONET RESEARCH

4X-0709.500 4X-0709.1 4X-0709.5 4X-0709.10
BIOT4X-0709.500EA 0 EUR
BIOT4X-0709.500 BIOT4X-0709.1 BIOT4X-0709.5 BIOT4X-0709.10
2-Piperidinonicotinonitril
2-Piperidinonicotinonitril
Formel: C₁₁H₁₃N₃
MDL: MFCD04113613
CAS-Nummer: 34595-23-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR