2-Piperidinoisonicotinonitril
Lieferant: BIONET RESEARCH
9X-0702.1
9X-0702.10
9X-0702.5
9X-0702.500
BIOT9X-0702.1EA
0
EUR
BIOT9X-0702.1
BIOT9X-0702.10
BIOT9X-0702.5
BIOT9X-0702.500
2-Piperidinoisonicotinonitril
2-Piperidinoisonicotinonitril
Formel:
C₁₁H₁₃N₃ |
MDL:
MFCD06200992 CAS-Nummer: 127680-89-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...