D(+)-Glucuronsäure-6,3-lacton 99+%

Lieferant: Thermo Scientific

Synonyme: (R)-2-((2S,3R,4S)-3,4-Dihydroxy-5-oxotetrahydrofuran-2-yl)-2-hydroxyacetaldehyde, D-(+)-Glucuronolactone

225920250 225921000 225925000
ACRO225920250EA 29.7 EUR
ACRO225920250 ACRO225921000 ACRO225925000
D(+)-Glucuronsäure-6,3-lacton 99+%
D(+)-Glucuronsäure-6,3-lacton
Formel: C₆H₈O₆
Molecular Weight: 176,12 g/mol
Schmelzpunkt: 170…176 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00135622
CAS-Nummer: 32449-92-6
EINECS: 251-053-3

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 170 to 176 °C
Loss on drying ≤0.5 % (105 °C, 3 h)
Solubility (5 % in water) Clear colorless to light yellow
Specific optical rotation +17.0° to +20.0° (20 °C, 589 nm) (c=1, water)
Titration excess NaOH ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR