D(+)-Glucuronsäure-6,3-lacton ≥99%

Lieferant: Thermo Fisher Scientific

Synonyme: (R)-2-((2S,3R,4S)-3,4-Dihydroxy-5-oxotetrahydrofuran-2-yl)-2-hydroxyacetaldehyde, D-(+)-Glucuronolactone

A15861.14 A15861.22 A15861.36
A15861.14EA 28.3 EUR
A15861.14 A15861.22 A15861.36
D(+)-Glucuronsäure-6,3-lacton ≥99%
D(+)-Glucuronsäure-6,3-lacton

Purity: 99% CAS No.: 32449-92-6 Molecular Formula: C6H8O6 Formula Weight: 176.12 Melting Point: 174° dec. Beilstein Reference: 83595 Merck Reference: 14,4467 Optical Rotation: +19° (c=8 in water) MDL No.: MFCD00135622

Formel: C₆H₈O₆
Molecular Weight: 176.12 g/mol
Schmelzpunkt: 174 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00135622
CAS-Nummer: 32449-92-6
EINECS: 251-053-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR