D(+)-Glucuronsäure-6,3-lacton

Lieferant: Serva

Synonyme: (R)-2-((2S,3R,4S)-3,4-Dihydroxy-5-oxotetrahydrofuran-2-yl)-2-hydroxyacetaldehyde, D-(+)-Glucuronolactone

22900.02 22900.03
SERA22900.02EA 0 EUR
SERA22900.02 SERA22900.03
D(+)-Glucuronsäure-6,3-lacton
D(+)-Glucuronsäure-6,3-lacton
Formel: C₆H₈O₆
Molecular Weight: 176,12 g/mol
MDL: MFCD00135622
CAS-Nummer: 32449-92-6
EINECS: 251-053-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR