D(+)-Glucuronsäure-6,3-lacton, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (R)-2-((2S,3R,4S)-3,4-Dihydroxy-5-oxotetrahydrofuran-2-yl)-2-hydroxyacetaldehyde, D-(+)-Glucuronolactone

M-Clarity™ Qualitätsstufe = MQ200

851450-100G-A 851450-1KG-A
SIAL851450-100G-AEA 73.6 EUR
SIAL851450-100G-A SIAL851450-1KG-A
D(+)-Glucuronsäure-6,3-lacton, Sigma-Aldrich®
D(+)-Glucuronsäure-6,3-lacton
Formel: C₆H₈O₆
Molecular Weight: 176.12 g/mol
MDL: MFCD00135622
CAS-Nummer: 32449-92-6
EINECS: 251-053-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR