Natrium-D(+)-glucuronat Monohydrat ≥99%
Lieferant: Thermo Fisher Scientific
Synonyme: D-Glucuronic acid sodium salt hydrate, D-Glucuronic acid sodium salt monohydrate, D(+)-Glucuronic acid, sodium salt, monohydrate, Sodium (2S,3S,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexanoate hydrate
L13985.06 L13985.14
L13985.06EA 24.1 EUR
L13985.06 L13985.14
Natrium-D(+)-glucuronat Monohydrat ≥99%
Natrium-D(+)-glucuronat Monohydrat
Purity: 99% CAS No.: 207300-70-7 Molecular Formula: C6H9NaO7.H2O Formula Weight: 234.14 (216.14anhy) Melting Point: 134-138° Beilstein Reference: 4218577
Formel: C₆H₁₀O₇·1H₂O
Molecular Weight: 234.14 g/mol (216.14 anhy)
Schmelzpunkt: 134…138 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD12966022
CAS-Nummer: 207300-70-7
EINECS: 239-065-7
Bestellen
TECHNISCHE DATEN
Environmentally Preferable