Hesperidin 95%
Lieferant: Thermo Scientific
Synonyme:
(S)-5-Hydroxy-2-(3-hydroxy-4-methoxyphenyl)-7-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-((((2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)methyl)tetrahydro-2H-pyran-2-yl)oxy)chroman-4-one
Purity: 95%
CAS No.: 520-26-3
Molecular Formula: C28H34O15
Formula Weight: 610.56
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Sensitivity: Hygroscopic
Appearance: White to pale brown
Melting Point: 250-255° dec.
Beilstein Reference: 75140
Merck Reference: 14,4671
MDL No.: MFCD00075663
Acts as an antioxidant
Formel:
C₂₈H₃₄O₁₅ Molecular Weight: 610,56 g/mol Schmelzpunkt: 250…255 °C (Zersetzung) |
MDL:
MFCD00075663 CAS-Nummer: 520-26-3 EINECS: 208-288-1 Merck Index: 14,4671 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...