Hesperidin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (S)-5-Hydroxy-2-(3-hydroxy-4-methoxyphenyl)-7-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-((((2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)methyl)tetrahydro-2H-pyran-2-yl)oxy)chroman-4-one

M-Clarity™ Qualitätsstufe = MQ200

H5254-100G H5254-25G
SIALH5254-100GEA 175 EUR
SIALH5254-100G SIALH5254-25G
Hesperidin, Sigma-Aldrich®
Hesperidin
Formel: C₂₈H₃₄O₁₅
MDL: MFCD00075663
CAS-Nummer: 520-26-3
EINECS: 208-288-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR