2-(2-Thienyliden)-3',4'-dimethoxyacetophenon

Lieferant: FLUOROCHEM

011223-25G 011223-100G
FLUO011223-25GEA 134 EUR
FLUO011223-25G FLUO011223-100G
2-(2-Thienyliden)-3',4'-dimethoxyacetophenon
2-(2-Thienyliden)-3',4'-dimethoxyacetophenon
Formel: C₁₅H₁₄O₃S
MDL: MFCD00170620
CAS-Nummer: 198639-89-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR