2-(2-Thienyliden)-3',4'-dimethoxyacetophenon

Lieferant: MENAI

CB789.100 CB789.25
MENACB789.100EA 0 EUR
MENACB789.100 MENACB789.25
2-(2-Thienyliden)-3',4'-dimethoxyacetophenon
2-(2-Thienyliden)-3',4'-dimethoxyacetophenon
Formel: C₁₅H₁₄O₃S
MDL: MFCD00170620
CAS-Nummer: 198639-89-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR