Aluminiummagnesiumisopropoxid 10% (w/v) in 2-Propanol , 99.9% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Magnesium aluminum i-propoxide
36591.AC
36591.AE
36591.ACEA
89.3
EUR
36591.AC
36591.AE
Aluminiummagnesiumisopropoxid 10% (w/v) in 2-Propanol , 99.9% (Metall-Basis)
Aluminiummagnesiumisopropoxid
Purity: 99.9%
CAS No.: 69207-83-6
Molecular Formula: Mg[Al(OCH(CH3)2)4]2
Formula Weight: 550.53
Physical Form: Liquid
Sensitivity: Moisture Sensitive
Appearance: Colourless
Boiling Point: 130-142°/2.0mm
UN No.: 1219
MDL No.: MFCD00070426
Formel:
C₂₄H₅₆Al₂MgO₈ Molecular Weight: 550.965 g/mol Siedepunkt: 130…142 °C (2.0 mmHg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00070426 CAS-Nummer: 69207-83-6 UN: 1219 ADR: 3 II |