N(α)-Boc-D-histidin 98

Lieferant: Thermo Fisher Scientific

Synonyme: Boc-D-His-OH

L08810.03 L08810.MD
L08810.03EA 75.9 EUR
L08810.03 L08810.MD
N(α)-Boc-D-histidin 98
N(α)-Boc-D-histidin

Purity: 98+% CAS No.: 50654-94-9 Molecular Formula: C11H17N3O4 Formula Weight: 255.27 Storage Temperature: 2°C to 8°C Melting Point: ca 197° dec. Beilstein Reference: 4196575 Optical Rotation: -26° (c=1 in methanol) MDL No.: MFCD00037851

Formel: C₁₁H₁₇N₃O₄
Molecular Weight: 255.3 g/mol
Schmelzpunkt: ∼ 197 °C (Zers.)
MDL: MFCD00037851
CAS-Nummer: 50654-94-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR