N(α)-Cbz-L-histidin ≥98%
Lieferant: Thermo Scientific
Synonyme:
Cbz-His-OH, N(α)-Benzyloxycarbonyl-L-histidine, N-Cbz-L-histidine, Z-His-OH
L09114.03
L09114.06
L09114.03EA
34.2
EUR
L09114.03
L09114.06
N(α)-Cbz-L-histidin ≥98%
N(α)-Cbz-L-histidin
Purity: 98+%
CAS No.: 14997-58-1
Molecular Formula: C14H15N3O4
Formula Weight: 289.29
Melting Point: ca 165° dec.
Beilstein Reference: 3561682
Formel:
C₁₄H₁₅N₃O₄ Molecular Weight: 289,3 g/mol Schmelzpunkt: ∼ 165 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00065960 CAS-Nummer: 14997-58-1 EINECS: 239-084-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...