Tetrahydropyran-4-essigsäure

Lieferant: COMBI-BLOCKS

Synonyme: Tetrahydropyranyl-4-acetic acid

AM-1005-1G AM-1005-5G
COBBAM-1005-1GEA 50.9 EUR
COBBAM-1005-1G COBBAM-1005-5G
Tetrahydropyran-4-essigsäure
Tetrahydropyran-4-essigsäure
Formel: C₇H₁₂O₃
Molecular Weight: 144.1700 g/mol
MDL: MFCD01631204
CAS-Nummer: 85064-61-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR