AmNA (Arachidonoyl-m-nitroanilin)

Lieferant: Cayman Chemical

90059-10 90059-5 90059-50 90059-25
CAYM90059-10EA 118 EUR
CAYM90059-10 CAYM90059-5 CAYM90059-50 CAYM90059-25
AmNA (Arachidonoyl-m-nitroanilin)
AmNA (Arachidonoyl-m-nitroanilin)
Formel: C₂₆H₃₆N₂O₃
CAS-Nummer: 1175954-87-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR