Lansoprazol
Lieferant: Cayman Chemical
Synonyme:
2-(((3-Methyl-4-(2,2,2-trifluoroethoxy)pyridin-2-yl)methyl)sulphinyl)-1H-benzo[d]imidazole
13627-500
13627-1
13627-5
13627-10
CAYM13627-500EA
52
EUR
CAYM13627-500
CAYM13627-1
CAYM13627-5
CAYM13627-10
Lansoprazol
Lansoprazol
Formel:
C₁₆H₁₄F₃N₃O₂S Molecular Weight: 369,36 g/mol |
MDL:
MFCD00866873 CAS-Nummer: 103577-45-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...