Bis(pinakolato)dibor
Lieferant: FLUOROCHEM
Synonyme:
4,4,4',4',5,5,5',5'-Octamethyl-2,2'-bi(1,3,2-dioxaborolane), 4,4,5,5-tetramethyl-2-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3,2-dioxaborolane
019250-5G
FL19250.20
019250.100G
019250-25G
019250-1G
FLUO019250-5GEA
0
EUR
FLUO019250-5G
CHMPFL19250.20
FLUO019250.100G
FLUO019250-25G
FLUO019250-1G
Bis(pinakolato)dibor
Bis(pinakolato)dibor
Formel:
C₁₂H₂₄B₂O₄ Molecular Weight: 253.94 g/mol |
MDL:
MFCD00799570 CAS-Nummer: 73183-34-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...