(Cyclopropylmethyl)triphenylphosphoniumbromid ≥98%
Lieferant: Thermo Scientific
Achtung
L01145.06 L01145.14
L01145.06EA 96.9 EUR
L01145.06 L01145.14
(Cyclopropylmethyl)triphenylphosphoniumbromid ≥98%
(Cyclopropylmethyl)triphenylphosphonium bromide
Purity: 98+% CAS No.: 14799-82-7 Molecular Formula: C22H22BrP Formula Weight: 397.29 Melting Point: 178-182° Beilstein Reference: 4068302
Formel: C₁₄H₂₉ClNP
Molecular Weight: 397,29 g/mol
Schmelzpunkt: 178…182 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00051878
CAS-Nummer: 14799-82-7
EINECS: 238-862-7