Theophyllin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1,3-Dimethyl-1H-purine-2,6(3H,9H)-dione, Theophylline

M-Clarity™ Qualitätsstufe = MQ200

T1633-50G T1633-1KG T1633-100G T1633-250G
SIALT1633-50GEA 35.9 EUR
SIALT1633-50G SIALT1633-1KG SIALT1633-100G SIALT1633-250G
Theophyllin, Sigma-Aldrich®
Theophyllin
Formel: C₇H₈N₄O₂
Molecular Weight: 180.16 g/mol
MDL: MFCD00079619
CAS-Nummer: 58-55-9
EINECS: 200-385-7
UN: 2811
ADR: 6.1 III

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR