(R)-3-(Boc-amino)piperidin

Lieferant: COMBI-BLOCKS

Synonyme: tert-Butyl N-[(3R)-piperidin-3-yl]carbamate, (R)-3-Boc-aminopiperidine, (R)-tert-Butyl piperidin-3-ylcarbamate

AM-1743-5G AM-1743-1G AM-1743-25G
COBBAM-1743-5GEA 0 EUR
COBBAM-1743-5G COBBAM-1743-1G COBBAM-1743-25G
(R)-3-(Boc-amino)piperidin
(R)-3-(Boc-amino)piperidin
Formel: C₁₀H₂₀N₂O₂
Molecular Weight: 200.28 g/mol
MDL: MFCD03093382
CAS-Nummer: 309956-78-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR