(S)-3-(Boc-amino)piperidin

Lieferant: COMBI-BLOCKS

AM-1742-5G AM-1742-1G
COBBAM-1742-5GEA 0 EUR
COBBAM-1742-5G COBBAM-1742-1G
(S)-3-(Boc-amino)piperidin
(S)-3-(Boc-amino)piperidin
Formel: C₁₀H₂₀N₂O₂
MDL: MFCD03093383
CAS-Nummer: 216854-23-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR