(±)-3-(Boc-amino)piperidin

Lieferant: CHEMPUR

Synonyme: tert-Butyl piperidin-3-ylcarbamate, 3-Aminopiperidine, 3-BOC protected

60-0011.1 60-0011.5
CHMP60-0011.1EA 0 EUR
CHMP60-0011.1 CHMP60-0011.5
(±)-3-(Boc-amino)piperidin
(±)-3-(Boc-amino)piperidin
Formel: C₁₀H₂₀N₂O₂
Molecular Weight: 200,28 g/mol
MDL: MFCD03839941
CAS-Nummer: 172603-05-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR