4-(Boc-amino)piperidin
Lieferant: FLUOROCHEM
Synonyme:
4-N-(tert-Butoxycarbonyl)aminopiperidine, tert-Butyl piperidin-4-ylcarbamate, tert-butyl N-(piperidin-4-yl)carbamate, 4-Boc-Aminopiperidine, 4-N-BOC-Aminopiperidine
011547-5G
011547-25G
FLUO011547-5GEA
51.8
EUR
FLUO011547-5G
FLUO011547-25G
4-(Boc-amino)piperidin
4-(Boc-amino)piperidin
Formel:
C₁₀H₂₀N₂O₂ |
MDL:
MFCD00798171 CAS-Nummer: 73874-95-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...