3-Amino-1-Boc-piperidin 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: (±)-3-Amino-1-Boc-piperidin

H52816.03 H52816.06
H52816.03EA 47.6 EUR
H52816.03 H52816.06
3-Amino-1-Boc-piperidin 97%
3-Amino-1-Boc-piperidin

Purity: 97% CAS No.: 184637-48-7 Molecular Formula: C10H20N2O2 Formula Weight: 200.28 MDL No.: MFCD01861219

Formel: C₁₀H₂₀N₂O₂
Molecular Weight: 200,28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01861219
CAS-Nummer: 184637-48-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR