4-Amino-1-Boc-piperidin 97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: tert-Butyl 4-Amino-1-piperidinecarboxylate, 4-Amino-1-tert-butoxycarbonylpiperidine, tert-Butyl 4-aminopiperidine-1-carboxylate, N-Boc-4-piperidineamine, tert-Butyl 4-amino-1-piperidinecarboxylate, 4-Amino-1-Boc-piperidine, 4-Amino-1-piperidinecarboxylic acid tert-Butyl Ester

H30779.03 H30779.06
H30779.03EA 58 EUR
H30779.03 H30779.06
4-Amino-1-Boc-piperidin 97%
4-Amino-1-Boc-piperidin

Purity: 97% CAS No.: 87120-72-7 Molecular Formula: C10H20N2O2 Formula Weight: 200.28 Melting Point: 44-48° UN No.: 3259 MDL No.: MFCD01076201

Formel: C₁₀H₂₀N₂O₂
Molecular Weight: 200,28 g/mol
Schmelzpunkt: 44…48 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01076201
CAS-Nummer: 87120-72-7
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR