4-Amino-1-Boc-piperidin
Lieferant: COMBI-BLOCKS
Synonyme:
tert-Butyl 4-Amino-1-piperidinecarboxylate, 4-Amino-1-tert-butoxycarbonylpiperidine, tert-Butyl 4-aminopiperidine-1-carboxylate, N-Boc-4-piperidineamine, tert-Butyl 4-amino-1-piperidinecarboxylate, 4-Amino-1-Boc-piperidine, 4-Amino-1-piperidinecarboxylic acid tert-Butyl Ester
AM-1626-5G
AM-1626-25G
AM-1626-1G
AM-1626-100G
COBBAM-1626-5GEA
0
EUR
COBBAM-1626-5G
COBBAM-1626-25G
COBBAM-1626-1G
COBBAM-1626-100G
4-Amino-1-Boc-piperidin
4-Amino-1-Boc-piperidin
Formel:
C₁₀H₂₀N₂O₂ Molecular Weight: 200,28 g/mol |
MDL:
MFCD01076201 CAS-Nummer: 87120-72-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...