4-Amino-1-Boc-piperidin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
tert-Butyl 4-Amino-1-piperidinecarboxylate, 4-Amino-1-tert-butoxycarbonylpiperidine, tert-Butyl 4-aminopiperidine-1-carboxylate, N-Boc-4-piperidineamine, tert-Butyl 4-amino-1-piperidinecarboxylate, 4-Amino-1-Boc-piperidine, 4-Amino-1-piperidinecarboxylic acid tert-Butyl Ester
M-Clarity™ Qualitätsstufe = MQ100
640042-1G
640042-5G
SIAL640042-1GEA
40.3
EUR
SIAL640042-1G
SIAL640042-5G
4-Amino-1-Boc-piperidin, Sigma-Aldrich®
4-Amino-1-Boc-piperidin
Formel:
C₁₀H₂₀N₂O₂ |
MDL:
MFCD01076201 CAS-Nummer: 87120-72-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...