1-Boc-3-(methylamino)piperidin

Lieferant: COMBI-BLOCKS

Synonyme: 3-(Methylamino)piperidine, N1-BOC protected, 1-Boc-3-methylaminopieridine

AM-1761-1G AM-1761-5G AM-1761-25G
COBBAM-1761-1GEA 46.2 EUR
COBBAM-1761-1G COBBAM-1761-5G COBBAM-1761-25G
1-Boc-3-(methylamino)piperidin
1-Boc-3-(methylamino)piperidin
Formel: C₁₁H₂₂N₂O₂
Molecular Weight: 214,31 g/mol
MDL: MFCD04114980
CAS-Nummer: 392331-89-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR