(S)-1-Boc-3-hydroxypiperidin ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(S)-tert-Butyl 3-hydroxypiperidine-1-carboxylate, (3S)-3-Hydroxypiperidine, N-BOC protected
H52771.03
H52771.06
H52771.03EA
62.5
EUR
H52771.03
H52771.06
(S)-1-Boc-3-hydroxypiperidin ≥97%
(S)-1-Boc-3-hydroxypiperidin
Purity: 97%
CAS No.: 143900-44-1
Molecular Formula: C10H19NO3
Formula Weight: 201.27
Flash Point:
Formel:
C₁₀H₁₉NO₃ Molecular Weight: 201,27 g/mol Schmelzpunkt: 46…50 °C Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115307 CAS-Nummer: 143900-44-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...