4-(Benzyloxy)-3,5-dimethylbenzaldehyde 95%

Lieferant: Thermo Scientific

Synonyme: 4-Benzyloxy-3,5-dimethylbenzaldehyde

B22067.03 B22067.06
B22067.03EA 39.5 EUR
B22067.03 B22067.06
4-(Benzyloxy)-3,5-dimethylbenzaldehyde 95%
4-(Benzyloxy)-3,5-dimethylbenzaldehyde

Purity: 95% CAS No.: 144896-51-5 Molecular Formula: C16H16O2 Formula Weight: 240.30 Flash Point: 110°(230°F) Refractive Index: 1.5860 Boiling Point: 162-163°/1mm MDL No.: MFCD00800683

Formel: C₁₆H₁₆O₂
Molecular Weight: 240,30 g/mol
Siedepunkt: 162…163 °C (1 mmHg)
Dichte: 1,10
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00800683
CAS-Nummer: 144896-51-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR