4-(Benzyloxy)-3,5-dimethylbenzaldehyde

Lieferant: CHEMPUR

Synonyme: 4-Benzyloxy-3,5-dimethylbenzaldehyde

BTB14060.250 BTB14060.1
CHMPBTB14060.250EA 0 EUR
CHMPBTB14060.250 MAYBBTB14060.1
4-(Benzyloxy)-3,5-dimethylbenzaldehyde
4-(Benzyloxy)-3,5-dimethylbenzaldehyde
Formel: C₁₆H₁₆O₂
Molecular Weight: 240,30 g/mol
MDL: MFCD00800683
CAS-Nummer: 144896-51-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR