Allantoin

Lieferant: Merck

Synonyme: 1-(2,5-Dioxoimidazolidin-4-yl)urea

1.01015.5001 1.01015.9025
1.01015.5001EA 0 EUR
1.01015.5001 1.01015.9025
Allantoin
Allantoin
Formel: C₄H₆N₄O₃
Molecular Weight: 158.12 g/mol
MDL: MFCD00005260
CAS-Nummer: 97-59-6
EINECS: 202-592-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR