1H,1H,2H,2H-Perfluoroctylmethyldichlorsilan

Lieferant: CHEMPUR SILANE

SILS13100.5 SILS13100.25
SILNSILS13100.5EA 0 EUR
SILNSILS13100.5 SILNSILS13100.25
1H,1H,2H,2H-Perfluoroctylmethyldichlorsilan
1H,1H,2H,2H-Perfluoroctylmethyldichlorsilan
Formel: C₉H₇Cl₂F₁₃Si
Molecular Weight: 461,13 g/mol
MDL: MFCD00042360
CAS-Nummer: 73609-36-6
EINECS: 277-551-0
UN: 2920
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR